
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22572-40-3
:
Formula:CH₃CH₂N=C=N(CH₂)₃N(CH₃)₃I
MDL Number:MFCD00011833
:297.17 g/mol
Melting Pt:90…92 °C
EINECS:245-095-1
:GHS07
Related Information
Frequently Bought Together