Barium iodide, ultra dry ≥99.999% (metals basis), beads -10 Mesh
Catalog # 47118.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:391.14 g/mol
Merck Index:14,980
Melting Pt:740 °C
Density:5.15
Storage Temperature:Ambient
EINECS:237-276-9
UN:1564
ADR:6.1,III
:ampouled under argon
:GHS07
CAS Number:13718-50-8
Formula:BaI₂
MDL Number:MFCD00003452