Order Entry
Italy
ContactUsLinkComponent
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Flammable
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Catalog # 46803.03
CAS Number:  
Flammable
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Catalog # 46803.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:92390-26-6
 :
Formula:C₁₈H₂₇ClRu
MDL Number:MFCD07369035
 :379.94 g/mol
Melting Pt:143…147 °C
ADR:4.3,II
 :GHS02