Zeolite Y, hydrogen
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1318-02-1
:
Formula:SiO₂:Al₂O₃
MDL Number:MFCD00132601
Storage Temperature:Ambient
EINECS:215-283-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together