Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD02091734
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together