Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Aminoadenosine ≥97%

2-Aminoadenosine ≥97%

2-Aminoadenosine ≥97%
Certificates
2-Aminoadenosine ≥97%
2-Aminoadenosine ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 2096-10-8 Molecular Formula: C10H14N6O4 Formula Weight: 282.26 Physical Form: Powder MDL No.: MFCD00053556

Synonyms:  2,6-Diamino-9-(beta-D-ribofuranosyl)purine, (2R,3R,4S,5R)-2-(2,6-Diamino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2,6-Diaminopurine Riboside
  • CAS Number:2096-10-8
  • Formula:C₁₀H₁₄N₆O₄
  • MDL Number:MFCD00053556
  • Molecular Weight:282.26 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information