Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Invar®, iron nickel
Invar®, iron nickel ≥99.9% (metals basis), powder -325 mesh
Invar®, iron nickel ≥99.9% (metals basis), powder -325 mesh
44600.36
:
Thermo Fisher Scientific
:
Invar®, iron nickel ≥99.9% (metals basis), powder -325 mesh
44600.36
:
Thermo Fisher Scientific
:
Pk:
500 g
Seq:
3645
Environmentally Preferable:
:
:
3089
:
GHS02
:
GHS08
:
GHS07
:
Ambient
:
Fe:Ni₆₅:₃₅%
:
MFCD01741928
:
4.1 II