

Cerium(IV) isopropoxide (37 - 45% Ce)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63007-83-0
:
Formula:C₁₂H₂₈CeO₄
MDL Number:MFCD00061483
Melting Pt:140 °C (dec.)
ADR:4.1,III
:376.468 g/mol
:GHS05
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together