Poly(vinylidene fluoride)
About this item
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
Notes: Incompatible with strong oxidizing agents.
CAS No.: 24937-79-9 Molecular Formula: (-CH2CF2-)n Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 155-160° MDL No.: MFCD00084470
- CAS Number:24937-79-9
- Formula:(-CH₂CF₂-)n
- MDL Number:MFCD00084470
- Melting Pt:155…160 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
Notes: Incompatible with strong oxidizing agents.
CAS No.: 24937-79-9 Molecular Formula: (-CH2CF2-)n Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 155-160° MDL No.: MFCD00084470
- CAS Number:24937-79-9
- Formula:(-CH₂CF₂-)n
- MDL Number:MFCD00084470
- Melting Pt:155…160 °C
- Storage Temperature:Ambient
Documents
Specifications for Product Family
Specification Test Results
- Appearance (Color):White
- Form:Powder
- Viscosity:Melt viscosity 23.50 to 29.50 Kpoise



