

Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99% (metals basis), Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20634-12-2
:
Formula:H₁₂N₆O₆Pt
MDL Number:MFCD00011624
:387.216 g/mol
Melting Pt:262 °C (dec.)
Storage Temperature:Ambient
EINECS:243-929-9
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together