Order Entry
Italy
ContactUsLinkComponent
Poly(styrene), dry, latex microsphere 1 micron
Poly(styrene), dry, latex microsphere 1 micron
Catalog # 43711.ME
CAS Number:  
Poly(styrene), dry, latex microsphere 1 micron
Catalog # 43711.ME
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Ambient
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450