
1,6-Bis(acryloyloxy)hexane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13048-33-4
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00008631
:226.27 g/mol
Density:1.010
Flash Pt:>110 °C (>230 °F)
:Stabilised with hydroquinone (HQ) 90 ppm
EINECS:235-921-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together