Order Entry
Italy
ContactUsLinkComponent
Boron trifluoridedimethyl sulphide complex, purified
Boron trifluoridedimethyl sulphide complex, purified
  42970.AE
 :  
Boron trifluoridedimethyl sulphide complex, purified
  42970.AE
 :  
  • Pk:
    100 mL
  • Seq:
    2440
 :
 :129.94 g/mol
 :1.235
 :8,I
 :Packaged under argon in resealable ChemSeal™ bottles.
 :2920
 :GHS02
 :GHS05
 :GHS07
 :Ambient
 :353-43-5
 :BF₃·S(CH₃)₂
 :MFCD00013193