

Tetrakis(dimethylamino)germanium(IV) ≥99% (metals basis)
Catalog # 42760.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:248.90 g/mol
Boiling Pt:203 °C
Melting Pt:14 °C
Density:1.07
Flash Pt:34 °C (93 °F)
UN:3129
ADR:4.3,II
:GHS02
:GHS05
Storage Temperature:Ambient
CAS Number:7344-40-3
Formula:C₈H₂₄GeN₄
MDL Number:MFCD01863476