Order Entry
Italy
ContactUsLinkComponent
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 75.0 micron
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 75.0 micron
Catalog # 42750.AA
CAS Number:  
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 75.0 micron
Catalog # 42750.AA
CAS Number:  

Specifications

  • Pk:
    5 mL

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Freezer
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450