Inserisci ordine
Italy
ContactUsLinkComponent
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.10 micron
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.10 micron
Catalog # 42712.4Y
CAS Number:  
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.10 micron
Catalog # 42712.4Y
CAS Number:  

Specifications

  • Conf:
    15 mL
  • Seq:
    2295

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Refrigerata
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450