Order Entry
Italy
ContactUsLinkComponent
Ground and waste water pollution standard solution 5, Specpure®
Ground and waste water pollution standard solution 5, Specpure®
Catalog # 42580.AE
CAS Number:  
Ground and waste water pollution standard solution 5, Specpure®
Catalog # 42580.AE
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    Ground & Waste Water Pollution Standard Solution 5
  • MDL No.:
    MFCD03792307
  • UN No.:
    3264

Specifications

About this item

For US EPA Clean Water Act.

  • 4 Analytes

UN No.: 3264
MDL No.: MFCD03792307