Order Entry
Italy
ContactUsLinkComponent
Ground and waste water pollution standard solution 4, Specpure®
Ground and waste water pollution standard solution 4, Specpure®
Catalog # 42579.AE
CAS Number:  
Ground and waste water pollution standard solution 4, Specpure®
Catalog # 42579.AE
CAS Number:  

Specifications

  • Matrix:
    2-5% HNO₃ + (CHOH)₂(COOH)₂
  • Pk:
    100 mL
  • Description:
    Ground & Waste Water Pollution Standard Solution 4
  • MDL No.:
    MFCD03792306
  • UN No.:
    3264

Specifications

About this item

UN No.: 3264
MDL No.: MFCD03792306