

Di-sec-butoxyaluminoxytriethoxysilane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68959-06-8
:
Formula:C₁₄H₃₃AlO₆Si
MDL Number:MFCD00053771
:352.48 g/mol
Boiling Pt:>200 °C
Density:1.00
Flash Pt:46 °C (115 °F)
Storage Temperature:Ambient
EINECS:273-392-6
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together