Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Hexachloroplatinic(IV) acid solution (20% Pt)

Toxic
Health
Corrosive
Hexachloroplatinic(IV) acid solution (20% Pt)
Certificates
Hexachloroplatinic(IV) acid solution (20% Pt)
Hexachloroplatinic(IV) acid solution (20% Pt)

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen hexachloroplatinate (IV), Dihydrogen hexachloroplatinate (IV)
  • CAS Number:16941-12-1
  • Formula:H₂Cl₆Pt
  • MDL Number:MFCD00011330
  • Molecular Weight:409.81 g/mol
  • Merck Index:14,7526
  • ADR:8,III
  •  :Density: 2.431 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:241-010-7
  • UN:3264
Product Family Options

Product Information