

Rubidium hydroxide hydrate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12026-05-0
:
Formula:H₃O₂Rb
MDL Number:MFCD00149835
:120.49 g/mol
Merck Index:14,8285
Storage Temperature:Ambient
EINECS:215-186-0
UN:2678
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together