


Zinc naphthenate 67% (10% Zn) in mineral spirits
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12001-85-3
:
Formula:2(C₁₁H₇O₂)·Zn
MDL Number:MFCD00147699
:319.71 g/mol
Density:1.01
Flash Pt:>37.8° (100 °F)
UN:1993
ADR:3,III
:GHS08
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together