Order Entry
Italy
ContactUsLinkComponent
Dihydrogen hexafluorozirconate 45% (w/w) in aqueous solution
Toxic
Corrosive
Dihydrogen hexafluorozirconate 45% (w/w) in aqueous solution
Catalog # 39462.A3
CAS Number:  
Toxic
Corrosive
Dihydrogen hexafluorozirconate 45% (w/w) in aqueous solution
Catalog # 39462.A3
CAS Number:  

Specifications

  • Pk:
    2 kg
  • Seq:
    4058

Specifications

About this item

 :
Density:1.51
EINECS:234-666-0
ADR:8,II
 :205.215 Da
UN:2922
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:12021-95-3
Formula:H₂ZrF₆
MDL Number:MFCD00082965