Order Entry
Italy
ContactUsLinkComponent
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
Irritant
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
Catalog # 39289.ME
CAS Number:  
Irritant
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
Catalog # 39289.ME
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :468.34 g/mol
 :Packaged under argon.
 :GHS07
Storage Temperature:Ambient
CAS Number:99326-34-8
Formula:C₁₇H₂₄F₃O₃RhS
MDL Number:MFCD00143752