Aluminum silicon slug ≥99.999% (metals basis), Puratronic®, Length: 12.7 mm (0.5 in), Ø 6.35 mm (0.25 in)
Catalog # 38492.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Storage Temperature:Ambient
EINECS:231-072-3
CAS Number:11145-30-5
MDL Number:MFCD00143595