Iridium 1% on activated carbon wetted with ca. 50% water, powder, reduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-88-5
:
Formula:Ir
MDL Number:MFCD00011062
:192.22 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together