Palladium 3% on activated carbon powder wetted with ca. 50% water, unreduced
Catalog # 38297.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:106.420 g/mol
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167