
Manganese(II) oxalate dihydrate (≥30% Mn)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6556-16-7
:
Formula:C₂H₆MnO₆
MDL Number:MFCD00150452
:181.002 g/mol
Merck Index:14,5734
Melting Pt:100 °C (dec.)
Density:2.453
EINECS:211-367-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together