

Tetraammineplatinum(II) nitrate solution (3 - 4% Pt) (w/w)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20634-12-2
:
Formula:H₁₂N₆O₆Pt
MDL Number:MFCD00011624
:387.216 g/mol
Density:1.05
Storage Temperature:Ambient
EINECS:243-929-9
UN:3218
ADR:5.1,III
:GHS01
:GHS03
Related Information
Frequently Bought Together