Anti-CYP1A1 Goat Polyclonal Antibody
Catalog # EVBIEB11172
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:cytochrome P450, family 1, subfamily A, polypeptide 1
- Antigen symbol:CYP1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Gene ID:1543
- Antigen synonyms:AHRR|CYP1|AHH|P450DX|P450-C|CP11|P1-450
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:CQEKQLDENANVQ
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Approx 85-90kDa band observed in lysates of cell lines A549 and MCF7 (calculated MW of 58.2kDa according to NP_000490.1). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources.
Type: Primary
Antigen: CYP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human