Anti-FABP10A Rabbit Polyclonal Antibody
ABGEAP20719A
: Abgent
Type: Primary
Antigen: FABP10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Protein A & Peptide affinity purification
- Antibody type:Primary
- Epitope:N-terminal
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Host:Rabbit
- Sequence:KLAKDVKPVTEIQQ
- Concentration:0.49
- Molecular weight:14.004 kDa
- Shipping temperature:4 °C
- Antigen name:Fatty Acid Binding Protein 10a, liver basic
- Cat. No.:ABGEAP20719A
- Reactivity:Zebrafish
- Gene ID:171481
- Antigen synonyms:L-FABP|Lb-FABP|Lfabp|Zf-FABP10|fabp10|z-L-BABP|zgc:103719|zgc:92741
- Immunogen:This (DANRE) fabp10a antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 30-63 amino acids from the N-terminal region of human (DANRE) fabp10a.
- Conjugation:Unconjugated
- Antigen symbol:FABP10A
- Storage buffer:pH 7,4
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



