Anti-BAD Rabbit Polyclonal Antibody
Catalog # ABGEAP3777H
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:BCL2-associated agonist of cell death
- Antigen symbol:BAD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:572
- Antigen synonyms:BCL2L8|BBC2
- Modification:phospho-Thr137
- Storage buffer:pH 7,4
- Molecular weight:18.392 kDa
- Sequence:PRPKSAGUATQMRQSC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.88
- Shipping temperature:4 °C
- Immunogen:This BAD Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T137 of human BAD.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human