Anti-ERBB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP3781T
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Erb-b2 receptor tyrosine kinase 2
- Antigen symbol:ERBB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:2064
- Antigen synonyms:NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
- Modification:phospho-Tyr1196
- Storage buffer:pH 7,4
- Molecular weight:137.91 kDa
- Sequence:GAVENPEJLTPQGGAC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.35
- Shipping temperature:4 °C
- Immunogen:This ERBB2 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y1196 of human ERBB2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human