Order Entry
Italy
ContactUsLinkComponent
 
Anti-IKBKB Rabbit Polyclonal Antibody
Catalog # ABGEAP3795A
Supplier:  Abgent
Anti-IKBKB Rabbit Polyclonal Antibody
Catalog # ABGEAP3795A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase beta
  • Antigen symbol:
    IKBKB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3551
  • Antigen synonyms:
    IKK-beta|IMD15|IKKB|IKK2|NFKBIKB
  • Modification:
    phospho-Ser733
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    86.564 kDa
  • Sequence:
    TVREQDQBFTALDWSC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.32
  • Shipping temperature:
    4 °C
  • Immunogen:
    This IKKB Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S733 of human IKKB.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: IKBKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human