Anti-IKBKB Rabbit Polyclonal Antibody
Catalog # ABGEAP20326C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase beta
- Antigen symbol:IKBKB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:3551
- Antigen synonyms:IKK-beta|IMD15|IKKB|IKK2|NFKBIKB
- Amino acid number:587 to 615
- Storage buffer:pH 7,4
- Molecular weight:86.564 kDa
- Sequence:EKKVRVIYTQLSKTC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.36
- Shipping temperature:4 °C
- Immunogen:This IKKB antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 587-615 amino acids from the Central region of human IKKB.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: IKBKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human