Anti-HDGFRP3 Rabbit Polyclonal Antibody
Catalog # ABGEAP17374C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:hepatoma-derived growth factor, related protein 3
- Antigen symbol:HDGFRP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:50810
- Antigen synonyms:HDGF-2|HDGF2|CGI-142|HRP-3
- Amino acid number:15 to 44
- Storage buffer:pH 7,4
- Molecular weight:22.62 kDa
- Sequence:DELPEGAVKPPANKYC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.49
- Shipping temperature:4 °C
- Immunogen:This HDGFRP3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 15-44 amino acids from the Central region of human HDGFRP3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: HDGFRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human