Order Entry
Italy
ContactUsLinkComponent
 
Anti-CACFD1 Rabbit Polyclonal Antibody
  ABGEAP17389B
 :  Abgent
Anti-CACFD1 Rabbit Polyclonal Antibody
  ABGEAP17389B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    calcium channel flower domain containing 1
  • Antigen symbol:
    CACFD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    11094
  • Antigen synonyms:
    C9orf7|D9S2135|FLOWER
  • Amino acid number:
    123 to 151
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    18.47 kDa
  • Sequence:
    SALGKKGDAISYARC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.6
  • Shipping temperature:
    4 °C
  • Immunogen:
    This C9orf7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 123-151 amino acids from the C-terminal region of human C9orf7.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: CACFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse