Anti-POLR3C Rabbit Polyclonal Antibody
ABGEAP17225C
: Abgent
- Antibody type:Primary
- Antigen name:Polymerase (RNA) III (DNA directed) polypeptide C (62kDa)
- Antigen symbol:POLR3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:10623
- Antigen synonyms:RPC62|RPC3
- Amino acid number:202 to 230
- Storage buffer:pH 7,4
- Molecular weight:60.612 kDa
- Sequence:RPKYTTDNKEPIPDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.45
- Shipping temperature:4 °C
- Immunogen:This POLR3C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 202-230 amino acids from the Central region of human POLR3C.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: POLR3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human