Order Entry
Italy
ContactUsLinkComponent
Anti-HOXA1A Rabbit Polyclonal Antibody
Anti-HOXA1A Rabbit Polyclonal Antibody
  ABGEAZB10012A
 :  Abgent
Anti-HOXA1A Rabbit Polyclonal Antibody
  ABGEAZB10012A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    homeobox A1a
  • Antigen symbol:
    HOXA1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Zebrafish
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    58051
  • Antigen synonyms:
    Hox-a1|hoxa3a|id:ibd3526|z-4
  • Amino acid number:
    192 to 219
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    35.737 kDa
  • Sequence:
    KRNPPKTGKAGEYC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.49
  • Shipping temperature:
    4 °C
  • Immunogen:
    This DANRE hoxa1a antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 192-219 amino acids from the Central region of DANRE hoxa1a.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: HOXA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Zebrafish