Order Entry
Italy
ContactUsLinkComponent
Cation exchange chromatography columns, HiTrap™ Capto™ S ImpAct
Cation exchange chromatography columns, HiTrap™ Capto™ S ImpAct
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, HiTrap™ Capto™ S ImpAct
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 29-4004-59
  • 17-3717-51
  • 17-3717-55
  • Binding capacity:
  • > 85 mg BSA/mL medium, > 90 mg lysozyme/mL medium, > 100 mg IgG/ mL medium
  • > 85 mg BSA/mL medium, > 90 mg lysozyme/mL medium, > 100 mg IgG/ mL medium
  • > 85 mg BSA/mL medium, > 90 mg lysozyme/mL medium, > 100 mg IgG/ mL medium
  • pH range:
  • 4 - 12
  • 4 - 12
  • 4 - 12
  • Storage:
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol

Specifications

Related Information

Frequently Bought Together