
3,3'-Dimethylnaphthidine indicator for metal titration, Supelco®
Catalog # 1.03122.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance of solution (10 g/l; ethanol):passes test
- Loss on drying (110 °C):≤ 1.0 %
- Suitability as indicator (for metal titration):passes test
- Suitability for bromate determination in water and drinking water for Pharo- or NOVA-spectrophotometer:passes test
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100
:312.42 g/mol
EINECS:236-069-0
:GHS07
Storage Temperature:Ambient
CAS Number:13138-48-2
Formula:C₂₂H₂₀N₂
MDL Number:MFCD00042706