Order Entry
Italy
ContactUsLinkComponent
Anti-Rab18 Rabbit Polyclonal Antibody
Anti-Rab18 Rabbit Polyclonal Antibody
Catalog # BSBTPB9791
Supplier:  Boster Bio
undefined
Anti-Rab18 Rabbit Polyclonal Antibody
Catalog # BSBTPB9791
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Rab18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    22931
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Ras-related protein Rab-18(RAB18) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: Rab18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat