Order Entry
Italy
ContactUsLinkComponent
S-(2-Aminoethyl)isothiouronium bromide hydrobromide for synthesis, Sigma-Aldrich®
S-(2-Aminoethyl)isothiouronium bromide hydrobromide for synthesis, Sigma-Aldrich®
Catalog # 8.20064.0010
Supplier:  Merck
CAS Number:  
S-(2-Aminoethyl)isothiouronium bromide hydrobromide for synthesis, Sigma-Aldrich®
Catalog # 8.20064.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    50 g/l (20 °C)
  • Melting Point:
    188 - 191 °C
  • Molar Mass:
    281.02 g/mol
  • Synonyms:
    AET
  • Hill Formula:
    C3H11Br2N3S
  • Chemical Formula:
    H2NCH2CH2SC(=NH)NH2 * 2HBr

Specification Test Results

About this item

 :
 :281.01 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-257-0
CAS Number:56-10-0
Formula:C₃H₁₁Br₂N₃S
MDL Number:MFCD00037011