Order Entry
Italy
ContactUsLinkComponent
1-Hydroxy-2-pyridinthione sodium salt 40% in water
Irritant
Environment
1-Hydroxy-2-pyridinthione sodium salt 40% in water
Catalog # 8.19054.0250
Supplier:  Merck
CAS Number:  
Irritant
Environment
1-Hydroxy-2-pyridinthione sodium salt 40% in water
Catalog # 8.19054.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) miscible in all proportions
  • Boiling Point:
    100 °C (1013 hPa)
  • Density:
    1.22 g/cm3 (20 °C)
  • pH:
    8.5 - 10.5 (H2O, 20 °C)
  • Storage temperature:
    +15...+25 °C

Specification Test Results

About this item

 :
 :149.143 g/mol
ADR:9,III
 :M-Clarity™ quality level = MQ200
UN:3082
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:15922-78-8
Formula:C₅H₄NNaOS