Order Entry
Italy
ContactUsLinkComponent
Anti-Proteasome 19S 10B Rabbit Polyclonal Antibody
Anti-Proteasome 19S 10B Rabbit Polyclonal Antibody
Catalog # BOSSBS-9346R
Supplier:  Bioss
Anti-Proteasome 19S 10B Rabbit Polyclonal Antibody
Catalog # BOSSBS-9346R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome 19S 10B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    5706
  • Antigen synonyms:
    26S protease regulatory subunit 10B|PSMC6|Proteasome 26S subunit ATPase 6|SUG2.|26S proteasome AAA-ATPase subunit RPT4|PRS10_HUMAN|Proteasome subunit p42|PRS10
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

The 26S proteasome is a large complex involved in the intracellular degradation of proteins in eukaryotes. Ubiquitination by E3 ubiquitin ligases targets proteins for degradation by this complex. The 26S proteasome plays an important role in the regulation of many biological processes. It is composed of over 30 different subunits and contains at least two copies of each subunit. Assembly of this large complex is ATP-dependent. Due to its size, it is fairly unstable and often disassociates into subcomplexes (including a 20S core and two 19S regulatory complexes). The 26s proteasome p42A (also known as Rpn7 in yeast and S10 in human) is one of at least nine, non-ATPase lid subunits of the 19S regulatory complex. It is important in the proper assembly and stability of the 26S proteasome. The 19S regulatory complex recognizes ubiquitinated proteins, removes the ubiquitin chains and translocates the proteins to the 20S core for degradation.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Proteasome 19S 10B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat