Order Entry
Italy
ContactUsLinkComponent
Anti-Interferon alpha 16 Rabbit Polyclonal Antibody
Anti-Interferon alpha 16 Rabbit Polyclonal Antibody
  BOSSBS-6330R
 :  Bioss
Anti-Interferon alpha 16 Rabbit Polyclonal Antibody
  BOSSBS-6330R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Interferon alpha 16
  • Antigen symbol:
    IFNA16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    3449
  • Antigen synonyms:
    N alpha N protein|Interferon|Interferon alpha WA|N16_HUMAN.|alpha 16
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

IFNA16 is produced by macrophages and has antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Interferon alpha 16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human