Anti-C7ORF69 Rabbit Polyclonal Antibody
Codice catalogo BOSSBS-5101R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Chromosome 7 open reading frame 69
- Antigen symbol:C7orf69
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:80099
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
MFAP3L is a membrane protein that is highly expressed in testis. It contains 1 Ig-like C2-type (immunoglobulin-like) domain and has three named isoforms produced by alternative splicing. Its function remains unknown.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: C7ORF69
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat