Order Entry
Italy
ContactUsLinkComponent
Anti-PKA C alpha+beta Rabbit Polyclonal Antibody
Anti-PKA C alpha+beta Rabbit Polyclonal Antibody
  BOSSBS-2371R
 :  Bioss
Anti-PKA C alpha+beta Rabbit Polyclonal Antibody
  BOSSBS-2371R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase C type alpha and beta
  • Antigen symbol:
    PRKCA + PRKCB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    81491
  • Antigen synonyms:
    Protein kinase C beta type|PKC-A|PRKACA|PKC-B|Protein kinase C alpha type|PRKC Pan|PKC-alpha|PRKCA|PKC-beta|PKCB|PRKCB1|PKCA
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. PKA beta (catalytic subunit) is a member of the Ser/Thr protein kinase family and is a catalytic subunit of AMPK.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PKA C alpha+beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat