Anti-HCM Rabbit Polyclonal Antibody
BOSSBS-15427R
: Bioss
- Antibody type:Primary
- Antigen symbol:HCM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:HCMT1a|KIAA0398|OTTHUMP00000162509|hCMT1c|MCES_HUMAN|hcm1p|hypothetical protein KIAA0396|mRNA cap guanine-N7 methyltransferase|hCMT1|mRNA (guanine N(7)) methyltransferase|Rnmt.|RNA (guanine 7) methyltransferase|mRNA (guanine 7)methyltransferase|RG7MT1|DKFZp686H1252|mRNA cap methyltransferase|RNA guanine 7 methyltransferase|hMet|EC 2.1.1.56|mRNA (guanine-N(7)-)-methyltransferase|MET
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: HCM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat