Order Entry
Italy
ContactUsLinkComponent
Anti-C20orf107 Rabbit Polyclonal Antibody
Anti-C20orf107 Rabbit Polyclonal Antibody
Catalog # BOSSBS-15090R
Supplier:  Bioss
Anti-C20orf107 Rabbit Polyclonal Antibody
Catalog # BOSSBS-15090R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 209, member B
  • Antigen symbol:
    FAM209B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    388799
  • Antigen synonyms:
    dJ1153D9.4|C20orf107
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf107 gene product has been provisionally designated C20orf107 pending further characterization.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: C20orf107
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human